siRNA Details
Gene Symbol: Pdgfrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026840.3 | NP_081116.3 | 2 | 239 | ||
GenBank mRNA | AK004179.1 | - | 2 | 131 | ||
AK081847.1 | - | 2 | 265 | |||
AK132625.1 | - | 2 | 240 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
