siRNA Details
Gene Symbol: Tmem53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285812.1 | NP_001272741.1 | 2 | 251 | ||
NM_001285814.1 | NP_001272743.1 | 3 | 316 | |||
NM_001285815.1 | NP_001272744.1 | 3 | 334 | |||
NM_026837.3 | NP_081113.1 | 2 | 230 | |||
NM_029589.1 | NP_083865.1 | 3 | 152 | |||
GenBank mRNA | AK004164.1 | - | 2 | 193 | ||
AK005104.1 | - | 2 | 137 | |||
AK005885.1 | - | 3 | 156 | |||
BC019937.1 | - | 2 | 156 | 4023311 | ||
BC020121.1 | - | 2 | 160 | 3991677 | ||
BC039805.1 | - | 1 | 535 | |||
BE647557.1 | - | 3 | 223 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107964, XM_006238665, FQ219156.
Gene Ontology Categories:
