siRNA Details
Gene Symbol: Lzic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026963.5 | NP_081239.1 | 3 | 879 | ||
GenBank mRNA | AK007657.1 | - | 3 | 368 | ||
AK146824.1 | - | 3 | 946 | |||
AK210899.1 | - | 3 | 209 | |||
DQ058008.1 | - | 2 | 204 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK161071.1 | - | 461 | Chr. - - [-] on Build |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
