siRNA Details
Gene Symbol: Krtap4-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027087.3 | NP_081363.1 | 1 | 647 | ||
GenBank mRNA | AK009035.1 | - | 1 | 648 |
Gene Symbol: Gm11554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099313.2 | NP_001092783.1 | 1 | 718 | ||
GenBank mRNA | BC104358.1 | - | 1 | 723 | 40049658 | |
BC104359.1 | - | 1 | 723 | 40049667 | ||
BC119472.1 | - | 1 | 723 | 40049663 | ||
BC119473.1 | - | 1 | 723 | 40049668 |
Gene Symbol: Gm11569 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099312.1 | NP_001092782.1 | 1 | 688 |
