siRNA Details
Gene Symbol: Zfp33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009327.2 | - | 1 | 195 | ||
AK049668.1 | - | 539 | ||||
BC046593.1 | - | 1 | 360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
