siRNA Details
Gene Symbol: Mif4gd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243584.1 | NP_001230513.1 | 3 | 331 | ||
NM_001243586.1 | NP_001230515.1 | 2 | 615 | |||
NM_001243587.1 | NP_001230516.1 | 3 | 331 | |||
NM_027162.4 | NP_081438.4 | 2 | 615 | |||
GenBank mRNA | AK003708.1 | - | 392 | |||
AK010175.1 | - | 3 | 285 | |||
AK150749.1 | - | 3 | 285 | |||
BC026740.1 | - | 3 | 291 | 4015105 | ||
CX210298.1 | - | 3 | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
