siRNA Details
Gene Symbol: Zmynd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252584.2 | NP_001239513.1 | 23 | 4612 | ||
NM_001252585.1 | NP_001239514.1 | 22 | 4504 | |||
NM_001281926.1 | NP_001268855.1 | 23 | 4511 | |||
NM_001291158.1 | NP_001278087.1 | 24 | 6777 | |||
NM_027230.5 | NP_081506.3 | 24 | 4855 | |||
GenBank mRNA | AK008125.1 | - | 1 | 642 | ||
AK139219.1 | - | 1 | 978 | |||
AK147620.1 | - | 23 | 4512 | |||
AK155864.1 | - | 23 | 4420 | |||
BC060508.1 | - | 1 | 857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
