siRNA Details
Gene Symbol: Ikbip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027078.2 | NP_081354.1 | 3 | 420 | ||
GenBank mRNA | AK006283.1 | - | 3 | 388 | ||
BC034890.1 | - | 3 | 382 | 1447362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
