siRNA Details
Gene Symbol: Taf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015889.2 | NP_001015889.1 | 3 | 669 | ||
NM_027139.5 | NP_081415.1 | 3 | 661 | |||
GenBank mRNA | AF305839.1 | - | 2 | 559 | ||
AK009322.1 | - | 1 | 409 | |||
BC042723.1 | - | 3 | 636 | 5006430 | ||
BC043028.1 | - | 2 | 493 | 5692221 | ||
BC096683.1 | - | 2 | 1409 | |||
CN837441.1 | - | 1 | 310 |

More Information
Other Species Targeted

Gene Ontology Categories:
