siRNA Details
Gene Symbol: MTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000254.2 | NP_000245.2 | 2 | 588 | ||
NM_001291939.1 | NP_001278868.1 | 2 | 588 | |||
NM_001291940.1 | NP_001278869.1 | 2 | 588 | |||
XM_005273141.4 | XP_005273198.1 | 2 | 557 | |||
GenBank mRNA | AK299904.1 | - | 2 | 588 | ||
AK307390.1 | - | 2 | 577 | |||
BC130616.1 | - | 2 | 187 | 40146647 | ||
BC136440.1 | - | 2 | 187 | 9020429 | ||
BC144095.1 | - | 2 | 187 | 9052615 | ||
BC144097.1 | - | 2 | 187 | 9052617 | ||
BC144098.1 | - | 2 | 187 | |||
U71285.1 | - | 2 | 228 | |||
U73338.1 | - | 2 | 559 | |||
U75743.1 | - | 2 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
