siRNA Details
Gene Symbol: Taf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015889.2 | NP_001015889.1 | 3 | 779 | ||
NM_027139.5 | NP_081415.1 | 3 | 771 | |||
GenBank mRNA | AF305839.1 | - | 2 | 669 | ||
AK009322.1 | - | 1 | 519 | |||
BC042723.1 | - | 3 | 746 | 5006430 | ||
BC043028.1 | - | 2 | 603 | 5692221 | ||
BC096683.1 | - | 2 | 1519 | |||
BC108348.1 | - | 3 | 701 | 3994683 | ||
BU962947.1 | - | 47 | ||||
CN837441.1 | - | 1 | 420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
