siRNA Details
Gene Symbol: 1600015I10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081273.2 | NP_001074742.2 | 4 | 2241 | ||
GenBank mRNA | AK005467.1 | - | 3 | 355 | ||
BC085280.1 | - | 4 | 2242 | |||
BC099440.1 | - | 4 | 878 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
