siRNA Details
Gene Symbol: Zmynd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252584.2 | NP_001239513.1 | 23 | 5013 | ||
NM_001252585.1 | NP_001239514.1 | 22 | 4905 | |||
NM_001281926.1 | NP_001268855.1 | 23 | 4912 | |||
NM_001291158.1 | NP_001278087.1 | 24 | 7178 | |||
NM_027230.5 | NP_081506.3 | 24 | 5256 | |||
GenBank mRNA | AK008125.1 | - | 1 | 1043 | ||
AK139219.1 | - | 1 | 1379 | |||
AK147620.1 | - | 23 | 4913 | |||
AK155864.1 | - | 23 | 4821 | |||
BB741303.1 | - | 1 | 233 | |||
BC012446.1 | - | 1 | 398 | |||
BC060508.1 | - | 1 | 1258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
