siRNA Details
Gene Symbol: Cdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013213.1 | - | 7 | 1163 | ||
AK147938.1 | - | 18 | 2244 | |||
AK167965.1 | - | 18 | 2244 | |||
AK168978.1 | - | 18 | 2216 | |||
BC005658.1 | - | 2 | 826 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
