siRNA Details
Gene Symbol: Rbm25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027349.3 | NP_081625.3 | 4 | 498 | ||
GenBank mRNA | AK011403.1 | - | 4 | 479 | ||
AK151910.1 | - | 4 | 450 | |||
AK162135.1 | - | 591 | ||||
AK164367.1 | - | 4 | 463 | |||
BC010792.1 | - | 3 | 244 | |||
BC023749.1 | - | 2 | 207 | |||
BC158104.1 | - | 3 | 369 | 9007388 | ||
CJ063347.1 | - | 4 | 491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
