siRNA Details
Gene Symbol: Gon4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242372.1 | NP_001229301.1 | 4546 | |||
NM_027389.3 | NP_081665.2 | 4495 | ||||
GenBank mRNA | AK122531.1 | - | 3867 | |||
AK147575.1 | - | 4497 | ||||
AK157887.1 | - | 3254 | ||||
AY040842.1 | - | 2458 | ||||
BC108377.1 | - | 20 | 3786 |
Gene Symbol: 5830417I10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017936.1 | - | 1 | 745 | ||
BC040255.1 | - | 1238 | ||||
BC059914.1 | - | 1 | 1315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
