siRNA Details
Gene Symbol: Mospd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290514.1 | NP_001277443.1 | 4 | 540 | ||
NM_027409.5 | NP_081685.1 | 4 | 540 | |||
GenBank mRNA | AK007533.1 | - | 4 | 473 | ||
AK088275.1 | - | 4 | 477 | |||
AK139795.1 | - | 4 | 484 | |||
BC018329.1 | - | 4 | 441 | 3962906 | ||
BC085100.1 | - | 4 | 448 | 6391017 | ||
BC100442.1 | - | 4 | 428 | 30501889 |
