siRNA Details
Gene Symbol: Atad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027435.2 | NP_081711.2 | 9 | 1174 | ||
GenBank mRNA | AK029867.1 | - | 2 | 321 | ||
AK032877.1 | - | 9 | 1175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
