siRNA Details
Gene Symbol: Tln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081242.2 | NP_001074711.2 | 38 | 5002 | ||
GenBank mRNA | AB093231.1 | - | 12 | 1544 | ||
AF467081.1 | - | 36 | 4731 | |||
AK029828.1 | - | 13 | 1695 | |||
BC033288.1 | - | 2 | 165 | |||
BC158057.1 | - | 38 | 4973 | 9007357 |

More Information
Other Species Targeted

Gene Ontology Categories:
