siRNA Details
Gene Symbol: Usp38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027554.2 | NP_081830.2 | 1 | 959 | ||
GenBank mRNA | AK028904.1 | - | 1 | 937 | ||
AK049287.1 | - | 1 | 927 | |||
AK052219.1 | - | 1 | 982 | |||
AK054116.1 | - | 1 | 961 | |||
AK129464.1 | - | 1 | 915 | |||
AK139639.1 | - | 1 | 972 | |||
AK142200.1 | - | 1 | 954 | |||
BC054404.1 | - | 1 | 959 | 6494448 | ||
BC057122.1 | - | 1 | 949 | 6834543 | ||
BC058784.1 | - | 1 | 887 | 6310341 | ||
BC066786.1 | - | 1 | 903 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
