siRNA Details
Gene Symbol: Krtap31-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025244.3 | NP_001020415.1 | 1 | 535 | ||
GenBank mRNA | AK037252.1 | - | 1 | 538 | ||
BC147120.1 | - | 1 | 503 | 9056124 | ||
BC147121.1 | - | 1 | 503 | 9056157 |
Gene Symbol: Krtap31-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027568.2 | NP_081844.1 | 1 | 535 | ||
GenBank mRNA | AK014635.1 | - | 1 | 536 | ||
BC147672.1 | - | 1 | 503 | 9056645 | ||
BC147678.1 | - | 1 | 503 | 9056651 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
