siRNA Details
Gene Symbol: 5830417I10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017936.1 | - | 1 | 748 | ||
BC040255.1 | - | 1241 | ||||
BC059914.1 | - | 1 | 1318 |
Gene Symbol: Gon4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242372.1 | NP_001229301.1 | 4549 | |||
NM_027389.3 | NP_081665.2 | 4498 | ||||
GenBank mRNA | AK122531.1 | - | 3870 | |||
AK147575.1 | - | 4500 | ||||
AK157887.1 | - | 3257 | ||||
AY040842.1 | - | 2461 | ||||
BC108377.1 | - | 20 | 3789 |

More Information
Other Species Targeted

Gene Ontology Categories:
