siRNA Details
Gene Symbol: Armc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027456.1 | NP_081732.1 | 3 | 230 | ||
NM_030184.2 | NP_084460.1 | 3 | 265 | |||
GenBank mRNA | AK014450.1 | - | 3 | 235 | ||
AK017564.1 | - | 3 | 231 | |||
AK019512.1 | - | 3 | 260 | |||
AK019600.1 | - | 3 | 254 | |||
AK030307.1 | - | 3 | 262 | |||
AK036083.1 | - | 3 | 236 | |||
AK039800.1 | - | 3 | 238 | |||
AK045699.1 | - | 3 | 229 | |||
AK046693.1 | - | 3 | 248 | |||
AK048064.1 | - | 3 | 244 | |||
AK140598.1 | - | 3 | 261 | |||
AK161393.1 | - | 3 | 245 | |||
AK164041.1 | - | 3 | 265 | |||
BC043101.1 | - | 3 | 217 | 6406321 |
