siRNA Details
Gene Symbol: Usp38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027554.2 | NP_081830.2 | 1 | 1036 | ||
GenBank mRNA | AK028904.1 | - | 1 | 1014 | ||
AK049287.1 | - | 1 | 1004 | |||
AK052219.1 | - | 1 | 1059 | |||
AK054116.1 | - | 1 | 1038 | |||
AK129464.1 | - | 1 | 992 | |||
AK139639.1 | - | 1 | 1049 | |||
AK142200.1 | - | 1 | 1031 | |||
BC054404.1 | - | 1 | 1036 | 6494448 | ||
BC057122.1 | - | 1 | 1026 | 6834543 | ||
BC058784.1 | - | 1 | 964 | 6310341 | ||
BC066786.1 | - | 1 | 980 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
