siRNA Details
Gene Symbol: Cfap43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027559.2 | NP_081835.2 | 4 | 654 | ||
GenBank mRNA | AK014581.1 | - | 672 | |||
AK015217.1 | - | 4 | 676 | |||
AK015497.1 | - | 4 | 655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
