siRNA Details
Gene Symbol: Spag9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199203.1 | NP_001186132.1 | 2 | 533 | ||
NM_001199204.1 | NP_001186133.1 | 2 | 533 | |||
NM_027569.2 | NP_081845.2 | 2 | 533 | |||
GenBank mRNA | AB047782.1 | - | 2 | 453 | ||
AF327451.1 | - | 2 | 515 | |||
AK030165.1 | - | 2 | 354 | |||
FJ042496.1 | - | 2 | 410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
