siRNA Details
Gene Symbol: Mllt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286158.1 | NP_001273087.1 | 6 | 1667 | ||
NM_027326.3 | NP_081602.3 | 11 | 3066 | |||
NM_029931.3 | NP_084207.2 | 7 | 1739 | |||
GenBank mRNA | AK019458.1 | - | 6 | 1659 | ||
AK136066.1 | - | 7 | 2300 | |||
AK147009.1 | - | 7 | 1653 | |||
BC021420.1 | - | 12 | 2902 | 5012671 |
