siRNA Details
Gene Symbol: Gon4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY040842.1 | - | 3827 |
Gene Symbol: 5830417I10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017936.1 | - | 4 | 1583 | ||
AK049055.1 | - | 3 | 607 | |||
BC040255.1 | - | 3610 | ||||
BC059914.1 | - | 3 | 3289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
