siRNA Details
Gene Symbol: Tln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081242.2 | NP_001074711.2 | 50 | 6730 | ||
GenBank mRNA | AB093231.1 | - | 24 | 3272 | ||
AF467081.1 | - | 46 | 6189 | |||
AK017597.1 | - | 4 | 502 | |||
AK029828.1 | - | 25 | 3423 | |||
AK040595.1 | - | 4 | 500 | |||
BC033288.1 | - | 12 | 1626 | |||
BC059856.1 | - | 4 | 491 | |||
BC079896.1 | - | 35 | 4593 | 30544295 | ||
BC158057.1 | - | 50 | 6701 | 9007357 |

More Information
Other Species Targeted

Gene Ontology Categories:
