siRNA Details
Gene Symbol: PHKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000293.2 | NP_000284.1 | 3, 4 | 365 | ||
NM_001031835.2 | NP_001027005.1 | 4, 5 | 477 | |||
XM_005255983.4 | XP_005256040.1 | 3, 4 | 328 | |||
XM_005255984.4 | XP_005256041.1 | 4, 5 | 628 | |||
XM_017023283.1 | XP_016878772.1 | 4, 5 | 450 | |||
XM_017023284.1 | XP_016878773.1 | 3, 4 | 331 | |||
GenBank mRNA | AK309226.1 | - | 3, 4 | 328 | ||
CR936710.1 | - | 4, 5 | 430 | |||
X84908.1 | - | 3, 4 | 337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
