siRNA Details
Gene Symbol: Spata16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027583.3 | NP_081859.1 | 10 | 1726 | ||
GenBank mRNA | AK014869.1 | - | 10 | 1690 | ||
BC100443.1 | - | 10 | 1707 | 30920480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
