siRNA Details
Gene Symbol: Cep170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099637.2 | NP_001093107.2 | 14, 15 | 4055 | ||
GenBank mRNA | AK016937.1 | - | 2, 3 | 316 | ||
AK038878.1 | - | 1, 2 | 594 | |||
AK172953.1 | - | 2, 3 | 1534 | |||
BC038377.1 | - | 2, 3 | 1486 | |||
BC085106.1 | - | 2, 3 | 303 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008769812, AB038495.
Gene Ontology Categories:
