siRNA Details
Gene Symbol: Cabyr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042418.1 | NP_001035883.1 | 4 | 779 | ||
NM_001042420.1 | NP_001035885.1 | 4 | 779 | |||
NM_027687.2 | NP_081963.1 | 4 | 779 | |||
GenBank mRNA | AF359382.1 | - | 4 | 779 | ||
AF359385.1 | - | 4 | 779 | |||
AK016856.1 | - | 4 | 786 | |||
AK132770.1 | - | 4 | 788 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
