siRNA Details
Gene Symbol: Lrrc72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177877.1 | NP_001171348.1 | 7 | 758 | ||
NM_027699.2 | NP_081975.1 | 3 | 255 | |||
GenBank mRNA | AK007140.1 | - | 3 | 256 | ||
AK016859.1 | - | 7 | 759 | |||
AK077182.1 | - | 3 | 633 | |||
BC107290.1 | - | 2 | 161 | 40053043 | ||
BC107291.1 | - | 2 | 161 | 40053045 |
