siRNA Details
Gene Symbol: Pds5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081321.1 | NP_001074790.1 | 16 | 1835 | ||
GenBank mRNA | AK172992.1 | - | 16 | 1875 | ||
BC021408.1 | - | 3 | 218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
