siRNA Details
Gene Symbol: Tns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289895.1 | NP_001276824.1 | 24 | 4079 | ||
NM_027884.3 | NP_082160.3 | 26 | 4140 | |||
GenBank mRNA | AK004758.1 | - | 3 | 667 | ||
BC052740.1 | - | 5 | 905 | |||
BC055076.1 | - | 3 | 717 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
