siRNA Details
Gene Symbol: Klhdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163729.2 | NP_001157201.2 | 11 | 1946 | ||
NM_027910.3 | NP_082186.2 | 12 | 2099 | |||
GenBank mRNA | AB053465.1 | - | 12 | 1970 | ||
AJ278127.1 | - | 12 | 1967 | |||
AK004964.1 | - | 12 | 2076 | |||
AK141673.1 | - | 11 | 1931 | |||
AK170623.1 | - | 12 | 1971 | |||
AK209958.1 | - | 1 | 75 | |||
BC018154.1 | - | 12 | 2067 | 3983436 | ||
BY606590.1 | - | 1 | 377 |
