siRNA Details
Gene Symbol: C530008M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163793.1 | NP_001157265.1 | 2 | 186 | ||
GenBank mRNA | AK015929.1 | - | 1 | 281 | ||
AK016430.1 | - | 3 | 496 | |||
AK030106.1 | - | 2 | 187 |

More Information
Other Species Targeted

Gene Ontology Categories:
