siRNA Details
Gene Symbol: Cep170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099637.2 | NP_001093107.2 | 15 | 4105 | ||
GenBank mRNA | AK016937.1 | - | 3 | 366 | ||
AK038878.1 | - | 2 | 644 | |||
AK172953.1 | - | 3 | 1584 | |||
BC038377.1 | - | 3 | 1536 | |||
BC085106.1 | - | 3 | 353 |

More Information
Other Species Targeted

Gene Ontology Categories:
