siRNA Details
Gene Symbol: Gpr39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027677.2 | NP_081953.2 | 2 | 1705 | ||
GenBank mRNA | AK016817.1 | - | 2 | 833 | ||
BC085285.1 | - | 2 | 1718 | 30745457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
