siRNA Details
Gene Symbol: Cabyr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042418.1 | NP_001035883.1 | 4 | 863 | ||
NM_001042420.1 | NP_001035885.1 | 4 | 863 | |||
NM_027687.2 | NP_081963.1 | 4 | 863 | |||
GenBank mRNA | AF359382.1 | - | 4 | 863 | ||
AF359385.1 | - | 4 | 863 | |||
AK016856.1 | - | 4 | 870 | |||
AK132770.1 | - | 4 | 872 |

More Information
Other Species Targeted

Gene Ontology Categories:
