siRNA Details
Gene Symbol: Ppp6r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027805.2 | NP_082081.1 | 8 | 1282 | ||
GenBank mRNA | AK018403.1 | - | 8 | 1201 | ||
AK084684.1 | - | 8 | 1283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
