siRNA Details
Gene Symbol: Pds5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081321.1 | NP_001074790.1 | 16 | 1861 | ||
GenBank mRNA | AK172992.1 | - | 16 | 1901 | ||
BC021408.1 | - | 3 | 244 |

More Information
Other Species Targeted

Gene Ontology Categories:
