siRNA Details
Gene Symbol: Speer4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027609.2 | NP_081885.1 | 2, 3 | 305 | ||
GenBank mRNA | AK015022.1 | - | 2, 3 | 308 | ||
BC116901.1 | - | 2, 3 | 331 | 40126220 | ||
BC116903.1 | - | 2, 3 | 331 | 40126222 |
