siRNA Details
Gene Symbol: Cep170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099637.2 | NP_001093107.2 | 19 | 4578 | ||
GenBank mRNA | AK016937.1 | - | 7 | 839 | ||
AK038878.1 | - | 6 | 1147 | |||
AK078541.1 | - | 2 | 60 | |||
AK172953.1 | - | 7 | 2087 | |||
BC038377.1 | - | 7 | 2039 | |||
BC085106.1 | - | 7 | 824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
