siRNA Details
Gene Symbol: Spata24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027733.5 | NP_082009.3 | 4 | 563 | ||
GenBank mRNA | AK016341.1 | - | 4 | 569 | ||
AY730532.1 | - | 4 | 563 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
