siRNA Details
Gene Symbol: 9030624J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027815.4 | NP_082091.3 | 7 | 1042 | ||
GenBank mRNA | AK011615.1 | - | 7 | 552 | ||
AK018573.1 | - | 7 | 582 | |||
AK035840.1 | - | 1 | 1655 | |||
AK050302.1 | - | 7 | 574 | |||
AK136140.1 | - | 7 | 582 | |||
BC054720.1 | - | 7 | 560 | 5689248 |

More Information
Other Species Targeted

Gene Ontology Categories:
