siRNA Details
Gene Symbol: Pds5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081321.1 | NP_001074790.1 | 22, 23 | 2657 | ||
GenBank mRNA | BC021408.1 | - | 9, 10 | 1040 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001083624, XM_008770153, XM_008770154, EF460313.
Gene Ontology Categories:
