siRNA Details
Gene Symbol: HPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000195.4 | NP_000186.2 | 12 | 1319 | ||
NM_001311345.1 | NP_001298274.1 | 11 | 1272 | |||
NM_001322476.1 | NP_001309405.1 | 12 | 1248 | |||
NM_001322477.1 | NP_001309406.1 | 12 | 1328 | |||
NM_001322478.1 | NP_001309407.1 | 11 | 1149 | |||
NM_001322479.1 | NP_001309408.1 | 11 | 1229 | |||
NM_001322480.1 | NP_001309409.1 | 10 | 987 | |||
NM_001322481.1 | NP_001309410.1 | 10 | 1067 | |||
NM_001322482.1 | NP_001309411.1 | 9 | 968 | |||
NM_001322483.1 | NP_001309412.1 | 11 | 1105 | |||
NM_001322484.1 | NP_001309413.1 | 11 | 1185 | |||
NM_001322485.1 | NP_001309414.1 | 10 | 1077 | |||
NM_001322487.1 | NP_001309416.1 | 12 | 1371 | |||
NM_001322489.1 | NP_001309418.1 | 10 | 1129 | |||
XM_005269757.4 | XP_005269814.1 | 12 | 1261 | |||
XM_017016170.1 | XP_016871659.1 | 9 | 875 | |||
XM_017016171.1 | XP_016871660.1 | 10 | 1082 | |||
XM_017016172.1 | XP_016871661.1 | 9 | 893 | |||
GenBank mRNA | U65676.1 | - | 12 | 1263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
