siRNA Details
Gene Symbol: Gm5800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034102.2 | NP_001029274.1 | 2 | 96 | ||
GenBank mRNA | BC049651.1 | - | 2 | 96 | ||
BC100479.1 | - | 2 | 96 | 30917737 |
Gene Symbol: 1700001F09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027940.2 | NP_082216.1 | 2 | 133 | ||
GenBank mRNA | AK005587.1 | - | 2 | 134 | ||
BC061060.1 | - | 2 | 115 | 6770724 |
Gene Symbol: 1700091H14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007033.1 | - | 2 | 256 |
Gene Symbol: BC061237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198677.1 | NP_941079.1 | 2 | 148 | ||
GenBank mRNA | BC061237.1 | - | 2 | 148 | 6705671 |
Gene Symbol: Gm10375 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098269.2 | NP_001091739.1 | 2 | 96 | ||
GenBank mRNA | BC089536.1 | - | 2 | 96 | 6742411 |
Gene Symbol: Gm10377 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244671.1 | NP_001231600.1 | 2 | 96 | ||
GenBank mRNA | BC049548.1 | - | 2 | 96 |
Gene Symbol: Gm17026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076616.1 | - | 2 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
